IKA topolino mobil Magnetic Stirrer without Heating; 1/Ea
Representative image only.
IKA topolino mobil Magnetic Stirrer without Heating; 1/Ea
IKA | Mfr: 3381300| Item: 1594942
For mixing of liquids independently of a mains power supply
Call For Price
Call For Price
Product Description
Includes
Plug-in power supply unit, four AA batteries
Key Features
  • Continually adjustable speed range up to 1800 rpm
  • Durable polypropylene top plate
  • 1 year warranty
More About this Item
This compact, low-profile mini stirrer is perfect for mixing quantities up to 250 mL. Its small size makes it convenient for use anywhere in the lab and maximizes premium benchtop space. Features durable brushless motor, high magnetic adhesion and optional power model for mains-free with standard battery operation or with supplied mains adapter without batteries or combination of both with batteries fitted.