IKA RH Basic 2 Magnetic Stirrer with Heating; 1/Ea
Representative image only.
IKA RH Basic 2 Magnetic Stirrer with Heating; 1/Ea
IKA | Mfr: 25006448| Item: 1635216
Economic magnetic stirrer with stainless steel heating plate
₹41,900.00INR / Each
₹41,900.00INR / Each
Product Description
Includes
Mains cable
Key Features
  • Fixed safety circuit 400 °C
  • Soft-start of the stirring motor prevents splashing or spraying
  • 1 year warranty
More About this Item
IKA RH basic 2 magnetic stirrer with heating function is suitable for tempering substances which are placed on the heating plate in vessels. By means of a built-in stirring drive and with the help of a magnetic bar within the vessel, the substances can be stirred and heated at the same time. The mixing intensity is dependent on the motor speed and on the size of the magnetic bar.