IKA RET Control-visc white Magnetic Stirrer with Heating, Enamel Plate; 1/Ea
Representative image only.
IKA RET Control-visc white Magnetic Stirrer with Heating, Enamel Plate; 1/Ea
IKA | Mfr: 5030000| Item: 1635222
First stirring hot plate to weigh, check pH, and monitor temperature
₹256,827.00INR / Each
₹256,827.00INR / Each
Product Description
Includes
External temperature sensor, Mains cable, Plastic screwdriver, H 104 protective cover, USB cable, magnetic stir bars (30 and 40 mm)
Key Features
  • Weigh and stir samples in one convenient unit
  • Monitor pH of your sample with built-in BNC connector
  • Built-in temperature sensor for precise temperature control
  • White ceramic-coated SS top for fast heating and excellent chemical resistance
  • 1 year warranty
More About this Item
This innovative unit offers the most advanced features within the market today, providing you the ability to monitor your sample activity more accurately. Monitor and weigh samples up to 2 kg using the integrated weighing function. Built-in BNC connector allows you to attach an electrode for analyzing and adjusting the pH of your sample. Set hot plate temperature up to 644°F (340°C) and observe the actual temperature of the sample with the temperature probe. Easily examine all parameters on the alphanumeric graphical TFT LCD with excellent image quality, easy-to-use navigation control, torque trend, and timer/countdown functions.
Round ceramic-coated stainless steel top provides quick and effective heat transfer to your samples. Improved heat control technology increases safety, while hot top indicator warns users to help prevent burns and injuries. Utilize the adjustable, from 122 to 698°F (50 to 380°C), over-temperature safety circuit to protect samples from overheating. Built-in RS-232 interface allows direct connection to a PC for control and documentation of trials for later study.