IKA RCT Basic package Magnetic Stirrer with Heating; 1/Ea
Representative image only.
IKA RCT Basic package Magnetic Stirrer with Heating; 1/Ea
IKA | Mfr: 9016422| Item: 1595379
Safety magnetic stirrer with heating, suitable for unsupervised operation
₹103,000.00INR / Each
₹103,000.00INR / Each
Product Description
Includes
RCT Basic magnetic stirrer, ETS-D5 electronic contact thermometer, support rod, holding rod and H 44 boss head clamp
Key Features
  • Strong motor for a higher range of speeds, additional temperature control mode for faster heating of medium
  • Exact temperature and speed setting via digital display, even when switched off
  • Set safety temperature limit displayed digitally
  • Hot Top indicator >> hot surface warning to prevent burns!
  • Digital error code display
  • 1 year warranty
More About this Item
RCT basic magnetic stirrer is a best seller in laboratories with integrated temperature control with adjustable safety circuit of heating plate temperature (50 to 360 °C). Bushing according to DIN 12878 for connecting a contact thermometer, e.g. ETS-D5, enables precise temperature control. High level of safety due to improved heat control technology. Enclosed assembly (IP42 rated) guarantees long service life. Round aluminium heating plate provides optimum heat transfer. Improved magnetic adhesion. Maximum stirring quantity (H20) is 20L with speed range of 50 to 1500 rpm and temperature range of ambient to 310 °C.