IKA Plate RCT Digital Magnetic Stirrer with Heating; 1/Ea
Representative image only.
IKA Plate RCT Digital Magnetic Stirrer with Heating; 1/Ea
IKA | Mfr: 25005229| Item: 1635221
Get temperature stability and high residual induction
₹105,000.00INR / Each
₹105,000.00INR / Each
Product Description
Includes
External temperature sensor PT 1000.60, Mains cable, Plastic screwdriver, USB cable, magnetic stir bars (30 and 40 mm)
Key Features
  • Stirs volumes up to 20 L and runs at maximum speed up to 1500 rpm
  • Independent control circuits for overtemperature protection
  • Hardened glass enclosure for spill and chemical resistance, safety and clear visibility
  • Integrated timer/counter for reminders and controlling sensitive reactions
  • 1 year warranty
More About this Item
The digital magnetic round-top stirring hot plate offers an integrated timer/counter for the control of kinetic sensitive reactions and reminders. The hardened glass enclosure design offers maximum visibility, chemical resistance, and safety. The use of an Alnico magnet provides excellent temperature stability, high residual induction, and maximum vortex.
Features intermittent and reverse direction modes for stirring.
Regular firmware updates via USB connection guarantee the hot plate will meet all your future needs. The stirrer can be operated by computer via an RS-232 or USB interface using the laboratory software labworldsoft®.