IKA myPlate Magnetic Stirrer without Heating; 1/Ea
Representative image only.
IKA myPlate Magnetic Stirrer without Heating; 1/Ea
IKA | Mfr: 25005156| Item: 1635233
Easily customize with your favourite picture
₹46,600.00INR / Each
₹46,600.00INR / Each
Product Description
Includes
power supply unit with exchangeable adapters
Key Features
  • Electronically controlled motor for strong stirring
  • Compact and accurate with digital LED speed display
  • 1 year warranty
More About this Item
This variable speed stirrer is ideal for low-viscosity fluids. Glass top plate offers excellent chemical resistance. Unit features digital LED speed display and speed can be varied between 100 to 2500 rpm.