IKA MVP 10 basic Membrane Vacuum Pump; 1/Ea
Representative image only.
IKA MVP 10 basic Membrane Vacuum Pump; 1/Ea
IKA | Mfr: 3980000| Item: 1635547
Diaphragm pump for dry, oil-free applications
₹237,773.00INR / Each
₹237,773.00INR / Each
Product Description
Includes
Power cord
Key Features
  • Slimline, compact design and long service life
  • Straightforward servicing and simple maintenance
  • Diaphragm pump that is highly resistant to chemicals as all parts that come into contact with gas are made of PTFE or PTFE compounds
More About this Item
Diaphragm vacuum pumps (also called membrane pumps) are oscillating positive displacement pumps. The diaphragms generate vacuum by expanding the volume in the vacuum chamber which in turn reduces the pressure in the vacuum chamber thus causing atmospheric pressure air to be pushed into the chamber. The air in this chamber is then pushed out from the vacuum chamber through a different route (gas outlet) with the use of valves. These periodic movements of the diaphragm generate vacuum at the inlet of the pump.