IKA KMO 3 Basic Magnetic Stirrer without Heating; 1/Ea
Representative image only.
IKA KMO 3 Basic Magnetic Stirrer without Heating; 1/Ea
IKA | Mfr: 20004631| Item: 1635234
Compact but powerful magnetic stirrer for mixing liquids
₹87,800.00INR / Each
₹87,800.00INR / Each
Product Description
Includes
Mains cable
Key Features
  • Reverse rotation for thorough mixing
  • USB interface for data acquisition
  • Illuminated set-up plate for clear visibility of sample mixing
  • 1 year warranty
More About this Item
This compact, powerful stirrer is perfect for mixing quantities up to 5L. Features clear white LED display for speed which can be varied up to 1500 rpm. Set-up plate is made of glass for better chemical resistance. Features key-lock button to avoid accidental parameter changes, reverse rotation for thorough mixing and integrated USB interface for data acquisition.