IKA H 135.50 Flask Carrier 2000 mL, without Handle, 7.7" Dia
Representative image only.
IKA H 135.50 Flask Carrier 2000 mL, without Handle, 7.7" Dia
IKA | Mfr: 0025000834| Item: 0467219
Perform multiple synthesis operations with these expansion blocks, flasks and inlays.
₹39,098.00INR / Each
₹39,098.00INR / Each
Product Description
Key Features
  • The aluminum blocks guarantee optimal heat transfer throughout the process without interference to the magnetic field.
  • Exchangeable flask inserts provide high-precision thermal conduction directly into the flask.
  • 2 year warranty
Two year warranty
Specifications
  • Compatible WithSynthesis Package 2
  • DescriptionFlask Carrier 2000 mL, without Handle, 7.7" Dia