IKA H 135.40 Flask Carrier 1000 mL, without Handle
Representative image only.
IKA H 135.40 Flask Carrier 1000 mL, without Handle
IKA | Mfr: 0025000833| Item: 0467217
Perform multiple synthesis operations with these expansion blocks, flasks and inlays.
₹32,077.00INR / Each
₹32,077.00INR / Each
Product Description
Key Features
  • The aluminum blocks guarantee optimal heat transfer throughout the process without interference to the magnetic field.
  • Exchangeable flask inserts provide high-precision thermal conduction directly into the flask.
  • 2 year warranty
Two year warranty
Specifications
  • Compatible WithSynthesis Package 2
  • DescriptionFlask Carrier 1000 mL, without Handle