IKA H 135.301 Flask Inlay 100 mL, 2.6" Inner Dia
Representative image only.
IKA H 135.301 Flask Inlay 100 mL, 2.6" Inner Dia
IKA | Mfr: 0025000641| Item: 0467214
Perform multiple synthesis operations with these expansion blocks, flasks and inlays.
₹10,241.00INR / Each
₹10,241.00INR / Each
Product Description
Key Features
  • The aluminum blocks guarantee optimal heat transfer throughout the process without interference to the magnetic field.
  • Exchangeable flask inserts provide high-precision thermal conduction directly into the flask.
  • 2 year warranty
Two year warranty
Specifications
  • Compatible WithSynthesis Package 2
  • DescriptionFlask Inlay 100 mL, 2.6" Inner Dia