IKA H 135.108 Block 250 mL, 3.1" x 3.1".
Representative image only.
IKA H 135.108 Block 250 mL, 3.1" x 3.1".
IKA | Mfr: 0025000633| Item: 0467208
Perform multiple synthesis operations with these expansion blocks, flasks and inlays.
₹9,178.00INR / Each
₹9,178.00INR / Each
Product Description
Key Features
  • The aluminum blocks guarantee optimal heat transfer throughout the process without interference to the magnetic field.
  • Exchangeable flask inserts provide high-precision thermal conduction directly into the flask.
  • 2 year warranty
Two year warranty
Specifications
  • Compatible WithSynthesis Package 1
  • DescriptionBlock 250 mL, 3.1" x 3.1".