IKA C-MAG HS 7 Digital Magnetic Stirrer with Heating, Ceramic Plate; 1/Ea
Representative image only.
IKA C-MAG HS 7 Digital Magnetic Stirrer with Heating, Ceramic Plate; 1/Ea
IKA | Mfr: 3487022| Item: 1594465
Precise temperature control with superior stirring strength
₹78,600.00INR / Each
₹78,600.00INR / Each
Product Description
Includes
external temperature sensor PT 1000, mains cable
Key Features
  • Precise temperature control through Pt 1000 temperature sensor
  • Stir up to 15 L volume (water) with a powerful motor
  • LCD display for exact temperature setting which shows set and actual temperature
  • 3 unique modes of operation guarantee optimal adjustment to your working method
  • Hot surface warning indicator prevents burns
  • Available stirring capacity: 5 L, 10 L, 15 L
  • 1 year warranty
More About this Item
Microprocessor-controlled stirring hot plates feature an over-temperature safety circuit fixed at 550°C. Easy to clean white ceramic top plate also provides excellent resistance to chemicals. Control panel is elevated in design to protect against accidental spillage. Units feature three unique modes of operation guarantee optimal adjustment to your working method: 1) Mode A is suitable for work with varying parameters and is safety temperature adjustable; 2) Mode B is suitable for series operation under uniform conditions; and 3) Mode C is suitable for unsupervised operation.