IKA 9024222 T-25 ULTRA-TURRAX Digital Homogenizer Kit
Representative image only.
IKA 9024222 T-25 ULTRA-TURRAX Digital Homogenizer Kit
IKA | Mfr: 9024222| Item: 1594377
Handle sample volumes from 0.001 to 2 litres
₹316,000.00INR / Each
₹316,000.00INR / Each
Product Description
Includes
stand with an 11-1/2"L x 7-5/8"W base and 24"L stainless steel rod, beaker/test tube holder, two movable clamps for positioning the shaft/generator and beaker/test tube holder, and a tool kit for shaft assembly. 10-ft power cord (the 115 VAC m
Key Features
  • System can be used with tough solids or large volume samples
  • Generator made of 316 stainless steel
  • 1 year warranty
More About this Item
This system is ideal for batch homogenizing of cell tissues, emulsifying suspended solids, and air pollution filter extraction. The key component is the rotor-stator generator. Shielded in a 316 stainless steel shaft, the rotor acts as a centrifugal pump to recirculate the liquid and suspended solids through the generator, where shear, impact, collision, and cavitation provide rapid homogenization.
1 Year Warranty
Specifications
  • DescriptionT-25 ULTRA-TURRAX Digital Homogenizer Kit