IKA 3378100 ETS-D6 contact Thermometer with pH
Representative image only.
IKA 3378100 ETS-D6 contact Thermometer with pH
IKA | Mfr: 0003378100| Item: 1594944
Safe for unsupervised operation
₹50,115.00INR / Each
₹50,115.00INR / Each
Product Description
Includes
1000Ω Pt external temperature sensor and 1.5-m power cord with European plug.
Key Features
  • Stronger motor for higher speed range — up to 1700 rpm
  • Temperature control mode for faster heating of medium
  • Platinum RTD temperature sensor ensures precise temperature control
More About this Item
Both units have a high safety level due to their improved heat control technology. You can digitally set safety temperature limit. Hot top indicator warns to prevent burns.
RCT basic Model features a polished aluminum round top plate which provides more efficient heat transfer than standard top plates. Hot plate features an over-temperature safety circuit adjustable from 122 to 590°F (50 to 310°C) to protect your samples from overheating. Adjustable speed up to 1500 rpm.
RET basic Model offers the same features of the RCT model plus a stainless steel top plate for better chemical resistance, higher temperatures, and a higher speed range up to 1700 rpm for more demanding applications.
Specifications
  • No. Of Stirring Positions1
  • DescriptionETS-D6 contact Thermometer with pH