IKA 3277700 Ultra-Flat Lab Disc Stirrer 230V
Representative image only.
IKA 3277700 Ultra-Flat Lab Disc Stirrer 230V
IKA | Mfr: 3277700| Item: 1594933
Ultra-flat design makes this ideal for use in hoods or cabinets
₹12,300.00INR / Each
₹12,300.00INR / Each
Product Description
Includes
cord with plug.
Key Features
  • Intrinsically safe
More About this Item
Stirrer utilizes an advanced magnetic coil technology with no moving parts, making the drive wear-free. Lab disc can be set up to reverse rotation direction every 30 sections to ensure thorough mixing.
Intrinsically Safe
Specifications
  • No. Of Stirring Positions1
  • DescriptionUltra-Flat Lab Disc Stirrer 230V