IKA 3240000 T 10 Basic Ultra-turrax Homogenizer
Representative image only.
IKA 3240000 T 10 Basic Ultra-turrax Homogenizer
IKA | Mfr: 3240000| Item: 1595052
125 Watt motor processes sample volumes of 0.5 to 100 mL up to 30,000 rpm
₹45,800.00INR / Each
₹45,800.00INR / Each
Buy It Together
Current Item
IKA 3240000 T 10 Basic Ultra-turrax Homogenizer
IKA 3240000 T 10 Basic Ultra-turrax Homogenizer
1595052
₹45,800.00INR / Each
+
IKA 3305500 S 10 N - 8 G Dispersing Element
1595057
₹142,000.00INR / Each
=
Total Price 187800.00 ADD 2 ITEMS TO CART
Product Description
Includes
storage case.
Key Features
  • Light weight and ergonomic form is ideal for manual operation
  • Quick-release coupling simplifies the removal of dispersing elements without the need for tools
  • Available disposable dispersion elements eliminate cross contamination -- great for PCR applications
More About this Item
These compact homogenisers are great for high-speed hand held operation. The 125-Watt motor offers stable operation in a variety of media samples, and variable speed control from 8,000 to 10,000 rpm. Disposable dispersion elements eliminate concerns of cross-contamination with DNA, or PCR analysis. Stainless steel dispersion elements remove quickly and clean completely for reuse. The rotor-stator generator uses a pump-like action to constantly draw in sample. Through a combination of shear, impact, collision, and cavitation, the generator rapidly disperses and disintegrates the material suspended in the media.
Specifications
  • DescriptionT 10 Basic Ultra-turrax Homogenizer