IKA 2953100 T50 Ultra-Turrax Homogeniser
Representative image only.
IKA 2953100 T50 Ultra-Turrax Homogeniser
IKA | Mfr: 2953100| Item: 1595054
700 Watt motor processes sample volumes of 0.25 to 30 L up to 10,000 rpm
₹149,000.00INR / Each
₹149,000.00INR / Each
Buy It Together
Current Item
IKA 2953100 T50 Ultra-Turrax Homogeniser
IKA 2953100 T50 Ultra-Turrax Homogeniser
1595054
₹149,000.00INR / Each
+ =
Total Price 455000.00 ADD 2 ITEMS TO CART
Product Description
Includes
homogenizer, support stand with clamp, and 2.4 m power cord with European plug.
Key Features
  • Variable speed control allows continuous operation
  • High-performance motor ensures reproducible operation even under changes in sample viscosity
  • Plug-in connectors simplify the removal of dispersing elements without the need for tools
More About this Item
This heavy-duty dispersing unit quickly Homogenises tissues and solids in batches from 0.25 to 30 liters. The rotor-stator generator uses a pump-like action to constantly draw in sample. Through a combination of shear, impact, collision, and cavitation, the generator rapidly disperses and disintegrates the material suspended in the media. Unit is designed for continuous operation and features an infinitely variable speed control from 4,000 to 10,000 rpm.
Conformite Europeenne
Specifications
  • DescriptionT50 Ultra-Turrax Homogeniser