IKA 2905200 Blade for Pulverizing Soft, Fibrous Materials, 440 SS
Representative image only.
IKA 2905200 Blade for Pulverizing Soft, Fibrous Materials, 440 SS
IKA | Mfr: 2905200| Item: 0430054
Various blade shapes and materials available to suit your application
₹22,400.00INR / Each
₹22,400.00INR / Each
Product Description
Key Features
  • For pulverizing soft, fibrous grinding materials
  • 2 year warranty
Two year warranty
Specifications
  • Material404 SS
  • Compatible WithHandheld analytical mills 04300-10 and -15
  • TypeOval shaped blade
  • DescriptionCutting blade, 440 SS for pulverizing soft, fibrous materials
  • Warranty2 Years