IKA 1443400 Blade for Crushing Fibrous Substances, Star-Shaped, 301 SS
Representative image only.
IKA 1443400 Blade for Crushing Fibrous Substances, Star-Shaped, 301 SS
IKA | Mfr: 0001443400| Item: 0430026
Various blade shapes and materials available to suit your application
₹24,200.00INR / Each
₹24,200.00INR / Each
Product Description
Key Features
  • Used to crush fibrous substances such as paper and vegetation, but also for plastics and material with a low specific weight
  • 3 month warranty
3 Month Warranty
Specifications
  • Material301 SS
  • Compatible WithHeavy-duty analytical mill 04301-10
  • TypeStar shaped blade
  • DescriptionStar-Shaped 301 SS Blade for the Heavy-Duty Analytical Mill
  • Warranty3 Months