IKA 0521800 Blade for Hardness of up to 9 Mohs, Tungsten Carbide
Representative image only.
IKA 0521800 Blade for Hardness of up to 9 Mohs, Tungsten Carbide
IKA | Mfr: 0000521800| Item: 0430024
Various blade shapes and materials available to suit your application
₹51,900.00INR / Each
₹51,900.00INR / Each
Product Description
Key Features
  • For hard materials up to Mohs hardness 9
  • 3 month warranty
3 Month Warranty
Specifications
  • MaterialTunsten carbide
  • Compatible WithHeavy-duty analytical mill 04301-10
  • TypeStraight blade
  • DescriptionTungsten-Carbide Blade for the Heavy-Duty Analytical Mill
  • Warranty3 Months