Idex Wrench, 1/16" or 1/8" OD Tubing, 1/4" and 5/16" Hex Ends; 1/EA
Representative image only.
Idex Wrench, 1/16" or 1/8" OD Tubing, 1/4" and 5/16" Hex Ends; 1/EA
Idex | Mfr: 6810| Item: 0201306
Use to tighten fittings on high-pressure valves
₹23,600.00INR / Each
₹23,600.00INR / Each
Product Description
Key Features
  • Fits over 1/16" or 1/8" OD tubing
  • Fits over 1/16" or 1/8" OD tubing
  • 1 month warranty
More About this Item
Use the double-ended socket wrench and spanner to tighten fittings on high-pressure valves. Slotted to fit over 1/16" and 1/8" OD tubing and has 1/4" and 5/16" internal hex ends. The "Z" shape provides ideal leverage for changing sample loops and fittings while keeping one end from restricting the use of the other.
1 Month Warranty
Specifications
  • Compatible With1/4" and 5/16" hex nuts
  • DescriptionIdex 6810 Wrench, 1/16" or 1/8" OD Tubing, 1/4" and 5/16" Hex Ends; 1/EA