Idex VHP Wrench, 1/4" Hex, 10 in-lbs; 1/EA
Representative image only.
Idex VHP Wrench, 1/4" Hex, 10 in-lbs; 1/EA
Idex | Mfr: VHP-1000| Item: 0202264
Designed to deliver a precise amount of torque to the fitting system
₹168,000.00INR / Each
₹168,000.00INR / Each
Product Description
Key Features
  • For use with very high pressure fittings
  • Durable tools
  • Easy-to-use
More About this Item
Let these fitting tools assist you with your connections. The wrenches are easy-to-use and aid to the tightening process.
Specifications
  • Compatible WithUse with 1/4" hex head fittings
  • DescriptionVHP Wrench, 1/4" Hex, 10 in-lbs; 1/EA