Idex VHP Union Body, ZDV, SS, 0.007" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
Representative image only.
Idex VHP Union Body, ZDV, SS, 0.007" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
Idex | Mfr: U-411-01| Item: 0202483
Zero dead volume unions ensure accurate analysis
₹12,200.00INR / Each
₹12,200.00INR / Each
Product Description
Key Features
  • Precision machined from 316 stainless steel to withstand high pressures
  • Securely join fluid pathway components
  • Holds tight connection even through vibration
  • VHP ZDV union body
  • 10-32 coned configuration
  • 0.007" thru-hole diameter
  • Swept volume of 13 nL
  • 1 year warranty
More About this Item
These Idex® VHP stainless steel ZDV unions provide a tight, secure connection in your chromatography fluid path.
1 Year Warranty
Specifications
  • Fitting MaterialStainless Steel
  • Number Of Ports2
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • DescriptionVHP Union Body, ZDV, SS, 0.007" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA