Idex VHP One-Piece Fitting, Black PK, 1/16" OD Tubing, 10-32 Coned; 1/EA
Representative image only.
Idex VHP One-Piece Fitting, Black PK, 1/16" OD Tubing, 10-32 Coned; 1/EA
Idex | Mfr: PK-120BLK| Item: 0202256
Fittings for very high pressure applications
₹2,900.00INR / Each
₹2,900.00INR / Each
Product Description
Key Features
  • PK material enables the use of capillary tubing in UHPLC Quick component replacement with minimal down time
  • 10-32 coned configuration
  • Requires 8.0 in-lb (0.90 N-m) of torque
  • For 1/16" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
The most demanding HPLC applications require fittings that will meet higher pressure requirements than standard chromatography fittings can accommodate. They are perfect for use in high-performance analytical systems. VHP PK Fittings are manufactured from a proprietary PEEK blend (PK), which allows use at higher pressures and temperatures.
1 Year Warranty
Specifications
  • Fitting MaterialPK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)10-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.588
  • ColorBlack
  • Max Temperature (° F)392
  • Max Temperature (° C)200
  • Max Operating Pressure (PSI)12000
  • DescriptionVHP One-Piece Fitting, Black PK, 1/16" OD Tubing, 10-32 Coned; 1/EA