Idex VHP MicroTight Union, True ZDV, PEEK/SS, 1/32" OD Tubing, 6-32 Coned; 1/EA
Representative image only.
Idex VHP MicroTight Union, True ZDV, PEEK/SS, 1/32" OD Tubing, 6-32 Coned; 1/EA
Idex | Mfr: UH-632| Item: 0202511
Maintain secure connections even under high pressure
₹30,900.00INR / Each
₹30,900.00INR / Each
Product Description
Includes
Two fittings PK-126 and one gauge plug P-553.
Key Features
  • 316 stainless steel body withstands high pressures Holds tight connection even through vibration
  • VHP true ZDV union assembly with 6-32 coned configuration for 1/32" OD tubing
  • Max pressure 15,000 psi (1034 bar)
  • 1 year warranty
More About this Item
VHP MicroTight® Unions join two pieces of tubing in a connection that will withstand higher pressures using trademarked MicroTight technology.
1 Year Warranty
Specifications
  • Fitting MaterialSS/PEEK
  • Number Of Ports2
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (InchShort)6-32 coned
  • Compression Size / Tube Od (Port 1) (in)1/32
  • ColorNatural
  • Max Operating Pressure (PSI)15000
  • DescriptionIdex UH-632 VHP MicroTight Union, True ZDV, PEEK/SS, 1/32" OD Tubing, 6-32 Coned; 1/EA