Idex VHP Adjust Torque Driver, 4 mm Hex, 2-12 in-lbs; 1/EA

Representative image only.

Idex VHP Adjust Torque Driver, 4 mm Hex, 2-12 in-lbs; 1/EA

Idex | Mfr: VHP-4000| Item: 0202280

Secure a leak-tight connection

₹152,000.00INR / Each

₹152,000.00INR / Each
Product Description

Key Features

  • Wrenches for tightening chromatography fittings
  • Externally adjustable torque setting from 2 to 12 in-lb (0.23 to 1.35 N.m)
  • 1 year warranty

More About this Item

Durable and reliable wrench set comes in handy for your chromatography needs.

1 Year Warranty
Specifications
  • MaterialMetal
  • Compatible WithExtender tools to torque driver
  • DescriptionVHP-4000 VHP Adjustable Torque Driver, 4 mm Hex, 2 to 12 in-lbs; 1/EA
  • Warranty1 year
Reviews
Q&A