Idex Valve, 6/4, MRA, SV, SS; 1/EA
Representative image only.
Idex Valve, 6/4, MRA, SV, SS; 1/EA
Idex | Mfr: MRA100-000| Item: 4271875
Automated valves for rapid, consistent flow switching
₹2,512,000.00INR / Each
₹2,512,000.00INR / Each
Buy It Together
Current Item
Idex Valve, 6/4, MRA, SV, SS; 1/EA
Idex Valve, 6/4, MRA, SV, SS; 1/EA
4271875
₹2,512,000.00INR / Each
+ =
Total Price 2810000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Control either manually or remotely, with software or instrument
  • Operate at higher torque levels or very small flow paths
  • 1 year warranty
More About this Item
Automated rotary shear valves are designed for applications that require fast, consistent switching between flow streams. Connect valves to your PC or instrument via USB, BCD, serial port, or contact closure.
Use switching valves to switch one or more flow paths to a different destination. Nano scale models 02013-61 and 02016-04 deliver flow rates less than 100 µL/min. Use large bore models when flow rates are greater than 10 mL/min.
This Mass Rate Attenuator 100-000 stainless steel valve is an active splitting device (6-position, 4-port) that delivers a small aliquot of an
HPLC stream into a Mass Spec stream through a stator and rotor seal.
1 Year Warranty
Specifications
  • Port Size (NPT(F))6-position, 4-port
  • Connection TypeAccepts 10-32 male threaded fittings
  • Max Pressure (PSI)1000
  • Max Pressure (bar)69
  • Flow Pattern6-position, 4-port
  • DescriptionMass Rate Attenuator 100-000 stainless steel valve is an active splitting device (6-position, 4-port).