Idex Vacuum Degasser, 4 Channel, 480 µL Per Channel, 2 mL/min; 100 to 240 VAC
Representative image only.
Idex Vacuum Degasser, 4 Channel, 480 µL Per Channel, 2 mL/min; 100 to 240 VAC
Idex | Mfr: 0001-6354| Item: 0201294
Degas multiple solvent lines at the same time
₹935,000.00INR / Each
₹935,000.00INR / Each
Product Description
Key Features
  • Remove dissolved gasses continuously from mobile phases
  • Inert flow path
  • Low volume and easy to prime
  • Four channels
  • 1 year warranty
More About this Item
These in-line full stand-alone degassing systems efficiently remove unwanted dissolved gases from mobile phases. Their unique design provides the highest level of reliable performance and makes helium sparging unnecessary. Optimal continuous operation is maintained by vacuum system monitoring.
1 Year Warranty
Specifications
  • Max Flow Rate (mL/min)2
  • Min Flow Rate Per Syringe (microL/min)480
  • Power (VAC)110 to 240
  • Number of Channels4
  • DescriptionVacuum Degasser, 4 Channel, 480 µL Per Channel, 2 mL/min; 100 to 240 VAC