Idex Tubing Cap, Black ETFE, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Tubing Cap, Black ETFE, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-755| Item: 0201438
Designed to effectively cap and seal off tubing
₹1,000.00INR / Each
₹1,000.00INR / Each
Product Description
Key Features
  • Securely cap off tubing
  • For 1/4-28 flat-bottom port
  • 1 month warranty
More About this Item
Caps are made of ETFE for increased strength and low gas permeability.
1 Month Warranty
Specifications
  • MaterialETFE
  • ColorBlack
  • Compatible With1/4-28 flat-bottom port
  • DescriptionP-755 Tubing Cap, Black ETFE, 1/4-28 Flat-Bottom; 1/EA