Idex Threaded Adapter with Fitting, PEEK, 1/8" ID, 5/16-24 Flat Bottom (F) to 1/4" NPT(M)
Representative image only.
Idex Threaded Adapter with Fitting, PEEK, 1/8" ID, 5/16-24 Flat Bottom (F) to 1/4" NPT(M)
Idex | Mfr: U-504| Item: 0202499
Maintain secure connections to 1/8" and 1/4" NPT ports even under high pressure
₹6,300.00INR / Each
₹6,300.00INR / Each
Product Description
Includes
One fitting 02020-59
Key Features
  • Securely join together fluid pathway components
  • Manufactured from PEEK polymer
  • Includes fittings
  • Included fitting accepts 3/16" OD tubing
  • 1/8" ID thru-hole diameter
  • 70.4 µL swept volume
  • 5/16-24 Flat Bottom (F) to 1/4" NPT(M)
  • 1 year warranty
More About this Item
Made from PEEK, these adapters are durable and have excellent chemical resistance. Wrap NPT threads with thread seal tape to ensure leak-free seal.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Number Of Ports2
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(F) (port 1) (InchShort)5/16-24 Flat Bottom
  • Compression Size / Tube Od (Port 1) (in)3/16
  • ColorNatural
  • Max Operating Pressure (PSI)500
  • Orifice Dimensions (in)1/8
  • DescriptionThreaded Adapter with Fitting, PEEK, 1/8" ID, 5/16-24 Flat Bottom (F) to 1/4" NPT(M)