Idex Threaded Adapter, PEEK, 0.080" ID, 1/4-28 Flat Bottom (F) to 5/16-24 Flat Bottom (F)
Representative image only.
Idex Threaded Adapter, PEEK, 0.080" ID, 1/4-28 Flat Bottom (F) to 5/16-24 Flat Bottom (F)
Idex | Mfr: P-135| Item: 0202430
Maintain secure connections even under high pressure
₹5,900.00INR / Each
₹5,900.00INR / Each
Buy It Together
Current Item
Idex Threaded Adapter, PEEK, 0.080" ID, 1/4-28 Flat Bottom (F) to 5/16-24 Flat Bottom (F)
Idex Threaded Adapter, PEEK, 0.080" ID, 1/4-28 Flat Bottom (F) to 5/16-24 Flat Bottom (F)
0202430
₹5,900.00INR / Each
+ =
Total Price 6870.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Securely join together fluid pathway components
  • Manufactured from inert polymers
  • Holds tight connection even through vibration
  • 0.080" ID thru-hole diameter
  • 4.1 µL swept volume
  • 1 year warranty
More About this Item
Made from PEEK or PCTFE, these adapters are durable and have excellent chemical resistance.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Sterilization MethodGamma Irradiation; Ethylene Oxide; Autoclave
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 1) (InchShort)1/4-28 Flat Bottom
  • Thread size UNF(F) (port 2) (InchShort)5/16-24 Flat Bottom
  • Flow PatternStraight
  • ColorNatural
  • Max Temperature (° C)125
  • Max Operating Pressure (PSI)1000
  • DescriptionThreaded Adapter, PEEK, 0.080" ID, 1/4-28 Flat Bottom (F) to 5/16-24 Flat Bottom (F)