Idex Swaging Tool for TinyTight Fittings, 6-40 MINSTAC Port; 1/EA
Representative image only.
Idex Swaging Tool for TinyTight Fittings, 6-40 MINSTAC Port; 1/EA
Idex | Mfr: M-150| Item: 0202536
Secure a leak-tight connection
₹16,300.00INR / Each
₹16,300.00INR / Each
Product Description
Key Features
  • Wrenches for tightening chromatography fittings
  • Use with TinyTight fittingswith 6-40 MINSTAC port
  • 1 year warranty
More About this Item
Durable and reliable wrench set comes in handy for your chromatography needs.
Place the tool in a vice then tighten tubing, fitting, and ferrule into the tool. Once removed, the swaged ferrule will be held in place.
1 Year Warranty
Specifications
  • MaterialSS
  • Compatible WithTinyTight fittings with 6-40 MINSTAC port
  • DescriptionSwaging Tool for TinyTight Fittings, 6-40 MINSTAC Port; 1/EA