Idex Super Flangeless™ Nut, Standard Knurl, Black PPS, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK
Representative image only.
Idex Super Flangeless™ Nut, Standard Knurl, Black PPS, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK
Idex | Mfr: P-217X| Item: 0202146
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
₹9,700.00INR / Pkg of 10
₹9,700.00INR / Pkg of 10
Product Description
Key Features
  • Eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Standard knurl
  • Black PEEK
  • For 1/32" or 1/16" OD tubing
  • 10/PK
  • 1 year warranty
More About this Item
Super Flangeless™ nuts combined with ferrules are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
Pressure rating of nut depends on ferrule used.
1 Year Warranty
Specifications
  • Fitting MaterialPPS
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/32
  • Thread Size ISO Metric (M) (Port 1) (mm)6
  • ColorBlack
  • DescriptionSuper Flangeless™ Nut, Standard Knurl, Black PPS, 1/32" or 1/16" OD Tubing, M6 Flat Bottom; 10/PK