Idex Super Flangeless™ Ferrule, Yellow ETFE; 1/EA
Representative image only.
Idex Super Flangeless™ Ferrule, Yellow ETFE; 1/EA
Idex | Mfr: P-258-01| Item: 0202167
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
₹1,600.00INR / Each
₹1,600.00INR / Each
Buy It Together
Current Item
Idex Super Flangeless™ Ferrule, Yellow ETFE; 1/EA
Idex Super Flangeless™ Ferrule, Yellow ETFE; 1/EA
0202167
₹1,600.00INR / Each
+
Cole-Parmer FEP Tubing, 1/16" ID x 1/8" OD; 50 Ft
5011915
₹13,300.00INR / Pkg of 1
=
Total Price 14900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • 1 year warranty
More About this Item
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
Minimum order 25 units.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • ColorYellow
  • DescriptionSuper Flangeless™ Ferrule, Yellow ETFE; 1/EA