Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
Representative image only.
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
Idex | Mfr: P-350X| Item: 0201384
Combine with Super Flangeless™ nut for a high pressure holding flat-bottom fitting assembly
₹11,800.00INR / Pkg of 10
₹11,800.00INR / Pkg of 10
Buy It Together
Current Item
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
Idex Super Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK
0201384
₹11,800.00INR / Pkg of 10
+
Cole-Parmer FEP Tubing, 1/16" ID x 1/8" OD; 50 Ft
5011915
₹13,300.00INR / Pkg of 1
=
Total Price 25100.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Lock ring eliminates tubing twist that can loosen connections Resistant to loosening through vibration
  • Max pressure of 2500 psi (172 bar)
  • For 1/8" OD tubing
  • Natural PEEK/316 SS ring
  • 10/PK
  • 1 month warranty
More About this Item
Super Flangeless™ ferrules combined with nuts are an excellent choice for tubing assemblies and will stay tight even through vibration. Ferrules come with lock ring to hold the ferrule in place, preventing the nuts from sliding off in tubing assemblies. The lock ring also allows tightening without twisting the tubing.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK/SS
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 1) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorNatural
  • Max Operating Pressure (PSI)2500
  • DescriptionSuper Flangeless™ Ferrule, Natural PEEK/SS Ring, 1/8" OD Tubing, 1/4-28 Flat Bottom; 10/PK