Idex Stator Face Assembly for Valve Model 7725, SS; 1/EA
Representative image only.
Idex Stator Face Assembly for Valve Model 7725, SS; 1/EA
Idex | Mfr: 7725-026| Item: 0201560
Prolong the life of your valves
₹68,300.00INR / Each
₹68,300.00INR / Each
Product Description
Key Features
  • Stators need replacement only if the ports or sealing surfaces become damaged
  • Use with valve model 7725
  • 1 year warranty
More About this Item
Optimize performance of your manual chromatography valves with these replacement stators.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible WithValve model 7725
  • DescriptionStator Face Assembly for Valve Model 7725, SS; 1/EA