Idex Standard Knurl Torque Tool, 4 in-lbs (0.45 N m); 1/EA

Representative image only.

Idex Standard Knurl Torque Tool, 4 in-lbs (0.45 N m); 1/EA

Idex | Mfr: P-1000| Item: 0202540

Secure a leak-tight connection

₹215,000.00INR / Each

₹215,000.00INR / Each
Product Description

Key Features

  • Wrenches for tightening chromatography fittings
  • Delivers a set torque of 4 in-lb (0.45 N.m)
  • 1 year warranty

More About this Item

Durable and reliable wrench set comes in handy for your chromatography needs.

1 Year Warranty
Specifications
  • MaterialSS/PCTFE
  • Compatible WithStandard knurl head
  • DescriptionStandard Knurl Torque Tool, 4 in-lbs (0.45 N m); 1/EA
  • Warranty1 year
Reviews
Q&A