Idex SealTight™ Ferrule, Black PEEK, 1/16" OD Tubing, 10-32 or M6 Coned; 10/PK
Representative image only.
Idex SealTight™ Ferrule, Black PEEK, 1/16" OD Tubing, 10-32 or M6 Coned; 10/PK
Idex | Mfr: F-192X| Item: 0201345
Slotted end on ferrule creates second compression zone for a tight seal
₹15,500.00INR / Pkg of 10
₹15,500.00INR / Pkg of 10
Product Description
Key Features
  • Constructed of durable PEEK
  • 10-32 coned or M6 coned, 10/PK
  • 1 month warranty
More About this Item
Optimize performance of your SealTight™ fittings with these replacement ferrules.
1 Month Warranty
Specifications
  • MaterialPEEK
  • ColorBlack
  • Compatible With10-32 or M6 coned, for 1/16" OD tubing
  • DescriptionF-192X SealTight™ Ferrule, Black PEEK, 1/16" OD Tubing, 10-32 or M6 Coned; 10/PK