Idex SealTight™ Ferrule, Black PEEK, 1/16" OD Tubing, 10-32 or M6 Coned; 1/EA
Representative image only.
Idex SealTight™ Ferrule, Black PEEK, 1/16" OD Tubing, 10-32 or M6 Coned; 1/EA
Idex | Mfr: F-192| Item: 0201899
Slotted end on ferrule creates second compression zone for a tight seal
₹1,700.00INR / Each
₹1,700.00INR / Each
Product Description
Key Features
  • Constructed of durable PEEK
  • 10-32 coned or M6 coned, 1/EA
  • 1 year warranty
More About this Item
Optimize performance of your SealTight™ fittings with these replacement ferrules.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorBlack
  • Compatible With10-32 or M6 coned, for 1/16" OD tubing
  • DescriptionSealTight™ Ferrule, Black PEEK, 1/16" OD Tubing, 10-32 or M6 Coned; 1/EA