Idex Rotor Seal for Valve Models 7000/7010/7010-087/7040, ETFE; 1/EA
Representative image only.
Idex Rotor Seal for Valve Models 7000/7010/7010-087/7040, ETFE; 1/EA
Idex | Mfr: 7010-071| Item: 0201547
Replace rotor seals for longer valve life
₹40,500.00INR / Each
₹40,500.00INR / Each
Product Description
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7000/7010/7010-087/7040
  • ETFE seal
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.
1 Year Warranty
Specifications
  • MaterialETFE
  • Compatible WithValve models 7000/7010/7010-087/7040
  • DescriptionRotor Seal for Valve Models 7000/7010/7010-087/7040, ETFE; 1/EA