Idex Rotor Seal for 7750/XX700-100, TZ
Representative image only.
Idex Rotor Seal for 7750/XX700-100, TZ
Idex | Mfr: 7750-050| Item: 4271808
Replace rotor seals for longer valve life
₹39,500.00INR / Each
₹39,500.00INR / Each
Product Description
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Use with valve models 7750 and XX700-100
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.
Replace rotor seals for longer valve life
1 Year Warranty
Specifications
  • MaterialETFE
  • Compatible WithValve models 7750/XX700-100
  • DescriptionIDEX 7750-050 Rotor Seal for 7750/XX700-100, TZ