Idex Rotor Seal for 7750, Polyimide
Representative image only.
Idex Rotor Seal for 7750, Polyimide
Idex | Mfr: 7750-016| Item: 4271805
Replace rotor seals for longer valve life
₹39,900.00INR / Each
₹39,900.00INR / Each
Product Description
Key Features
  • Polymeric disc ensures a high pressure seal against the stator
  • Replace rotor seals for longer valve life
  • Polymeric disc ensures a high pressure seal against the stator
  • 1 year warranty
More About this Item
Vespel® offers high heat resistance. Biocompatible PEEK is inert to almost all organic solvents. ETFE provides excellent solvent resistance and good thread strength.
1 Year Warranty
Specifications
  • MaterialPolyimide
  • Compatible WithValve model 7750
  • DescriptionRotor Seal is a polymeric disc that ensures a high pressure seal against the stator.