Idex Ring Stand Valve Bracket
Representative image only.
Idex Ring Stand Valve Bracket
Idex | Mfr: 7160-029| Item: 0202521
Optimize performance of your Idex® valve
₹18,200.00INR / Each
₹18,200.00INR / Each
Product Description
Key Features
  • Mounting brackets and plates provide enhanced support for valves
  • Has 1.5 cm dia rod hole
  • 1 year warranty
More About this Item
Use mounting brackets and plates to provide steady support for valves.
The ring stand mounting bracket allows the valves to mount onto common laboratory equipment.
1 Year Warranty
Specifications
  • Compatible WithValves to mount on laboratory equipment
  • DescriptionIdex Ring Stand Valve Bracket; 1/EA