Idex RheBuild® Kit for Valve Models 9010/9040; 1/EA
Representative image only.
Idex RheBuild® Kit for Valve Models 9010/9040; 1/EA
Idex | Mfr: 9010-999| Item: 0201577
Regular maintenance ensures optimal valve performance
₹62,900.00INR / Each
₹62,900.00INR / Each
Product Description
Key Features
  • Optimize performance of your manual chromatography valves
  • Use with valve models 9010/9040
  • 1 year warranty
More About this Item
RheBulid® kits contains normal wear parts, tools for disassembly/reassembly, and instructions to maintain your valves.
1 Year Warranty
Specifications
  • Compatible WithValve models 9010/9040
  • DescriptionRheBuild® Kit for Valve Models 9010/9040; 1/EA