Idex RheBuild® Kit for Valve Model 7750E06
Representative image only.
Idex RheBuild® Kit for Valve Model 7750E06
Idex | Mfr: 7750-999| Item: 0203256
Cost-effective kits contain parts and tools
₹95,100.00INR / Each
₹95,100.00INR / Each
Product Description
Includes
Parts, tools and instructions.
Key Features
  • Contains those parts most likely to wear or get damaged
  • Eliminates the need to order individual parts
  • Maintaining valves for model 7750E06
  • 1 year warranty
More About this Item
These kits include all the necessary components you need to maintain your valves and injectors.
1 Year Warranty
Specifications
  • Compatible WithValve Model 7750E06
  • DescriptionRheBuild® Kit