Idex RheBuild® Kit for Actuated Valve 02013-62; 1/EA
Representative image only.
Idex RheBuild® Kit for Actuated Valve 02013-62; 1/EA
Idex | Mfr: 7150-999| Item: 0201309
Regular maintenance ensures optimal valve performance
₹197,000.00INR / Each
₹197,000.00INR / Each
Product Description
Includes
Two RPC-10 rotor seals
Key Features
  • Optimize performance of your rotary shear valves
  • Use with valve 02013-62
  • 1 month warranty
More About this Item
Routine valve maintenance using RheBuild® kits or rapid replacement pods ensure optimal performance. The entire pod can be replaced quickly as a unit with virtually no instrument downtime for repair.
1 Month Warranty
Specifications
  • Compatible WithActuated valve 02013-62
  • DescriptionRheBuild® Kit for Actuated Valve 02013-62; 1/EA