Idex RheBuild® Kit for 8 Models with 2 position, 6-port motorized switching injection valves
Representative image only.
Idex RheBuild® Kit for 8 Models with 2 position, 6-port motorized switching injection valves
Idex | Mfr: 7900-999| Item: 0203258
Cost-effective kits contain parts and tools
₹87,800.00INR / Each
₹87,800.00INR / Each
Product Description
Includes
Parts, tools and instructions.
Key Features
  • Contains those parts most likely to wear or get damaged
  • Eliminates the need to order individual parts
  • Maintaining valves for models MXP9900-000, MHP9900-500-1, MHP9900-000, PD9900, MX7900-000, MX9900-000, MX7925-000, MX9925-000
  • 1 year warranty
More About this Item
These kits include all the necessary components you need to maintain your valves and injectors.
1 Year Warranty
Specifications
  • Compatible WithMXP9900-000, MHP9900-500-1, MHP9900-000, PD9900, MX7900-000, MX9900-000, MX7925-000, MX9925-000
  • DescriptionRheBuild® Kit for 8 models with 2 position, 6-port motorized switching injection valves