Idex Rapid Replacement Pod for Actuated Valve 02016-05; 1/EA
Representative image only.
Idex Rapid Replacement Pod for Actuated Valve 02016-05; 1/EA
Idex | Mfr: PD9900| Item: 0201628
Regular maintenance ensures optimal valve performance
₹186,000.00INR / Each
₹186,000.00INR / Each
Product Description
Key Features
  • Optimize performance of your rotary shear valves
  • Use with valve 02016-05
  • 1 year warranty
More About this Item
Routine valve maintenance using RheBuild® kits or rapid replacement pods ensure optimal performance. The entire pod can be replaced quickly as a unit with virtually no instrument downtime for repair.
1 Year Warranty
Specifications
  • Compatible WithActuated valve 02016-05
  • DescriptionRapid Replacement Pod for Actuated Valve 02016-05; 1/EA