Idex Port Plug, Standard Knurl, Natural PEEK, 10-32 Coned Extra Long Thread ; 1/EA
Representative image only.
Idex Port Plug, Standard Knurl, Natural PEEK, 10-32 Coned Extra Long Thread ; 1/EA
Idex | Mfr: P-550| Item: 0201840
Available in various thread sizes and materials
₹1,000.00INR / Each
₹1,000.00INR / Each
Buy It Together
Current Item
Idex Port Plug, Standard Knurl, Natural PEEK, 10-32 Coned Extra Long Thread ; 1/EA
Idex Port Plug, Standard Knurl, Natural PEEK, 10-32 Coned Extra Long Thread ; 1/EA
0201840
₹1,000.00INR / Each
+ =
Total Price 1640.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easily identify columns with color coded plugs
  • For 10-32 coned port
  • Extra long thread
  • Natural PEEK construction
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors. Select coned plugs for high-pressure applications; flat-bottom plugs for low-pressure applications. Color-coded plugs are ideal for easily identifying stored columns that have different packing materials.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorNatural
  • Compatible With10-32 coned port
  • DescriptionPort Plug, Standard Knurl, Natural PEEK, 10-32 Coned Extra Long Thread; 1/EA